Learn More
MSC GF Gage Threaded Ring Gage, #2-64 Thread, Class 2A, Go & No Go
Catalog No. 5000530673
Encompass
This GF Gage threaded ring gage provides precise inspection for #2-64 threads with a Class 2A fit. Its double-ring design integrates both Go and No Go elements, making it suitable for rigorous quality control in metalworking and manufacturing environments.
  • Designed for #2-64 thread specifications
  • Features Class 2A thread classification
  • Includes both Go and No Go measuring elements
  • Utilizes a double ring configuration for comprehensive inspection
  • Ensures accurate thread inspection
Catalog No. 50-005-30673 Supplier MSC Supplier No. 55324990
This GF Gage threaded ring gage provides precise inspection for #2-64 threads with a Class 2A fit. Its double-ring design integrates both Go and No Go elements, making it suitable for rigorous quality control in metalworking and manufacturing environments.
  • Designed for #2-64 thread specifications
  • Features Class 2A thread classification
  • Includes both Go and No Go measuring elements
  • Utilizes a double ring configuration for comprehensive inspection
  • Ensures accurate thread inspection