Learn More
MSC GF Gage Threaded Ring Gage, 3/4-10 Thread, Class 2A, No Go, Single Ring
Catalog No. 5000528786
Encompass
This threaded ring gage is designed for inspecting external threads. It features a precise 3/4-10 thread size and is classified as a No Go gage, ensuring that components meet the specified Class 2A tolerance.
  • No Go type
  • Thread size 3/4-10
  • Class 2A classification
  • Single ring configuration
Catalog No. 50-005-28786 Supplier MSC Supplier No. 55323398
This threaded ring gage is designed for inspecting external threads. It features a precise 3/4-10 thread size and is classified as a No Go gage, ensuring that components meet the specified Class 2A tolerance.
  • No Go type
  • Thread size 3/4-10
  • Class 2A classification
  • Single ring configuration