Learn More
MSC GF Gage Threaded Ring Gage, 3/4-10 UNC, Class 3A, Go & No Go
Catalog No. 5000511514
Encompass
This product is a threaded ring gage, designed for precise measurement of threads. It features a 3/4-10 UNC thread type with a Class 3A classification. The set includes both Go and No Go gages, providing comprehensive inspection capabilities for threaded components.
  • Designed for accurate thread measurement.
  • Features a 3/4-10 UNC thread size and type.
  • Classified as Class 3A for precision applications.
  • Includes both Go and No Go gages for complete inspection.
  • Comes with a standard Certificate of Compliance.
Catalog No. 50-005-11514 Supplier MSC Supplier No. 55326870
This product is a threaded ring gage, designed for precise measurement of threads. It features a 3/4-10 UNC thread type with a Class 3A classification. The set includes both Go and No Go gages, providing comprehensive inspection capabilities for threaded components.
  • Designed for accurate thread measurement.
  • Features a 3/4-10 UNC thread size and type.
  • Classified as Class 3A for precision applications.
  • Includes both Go and No Go gages for complete inspection.
  • Comes with a standard Certificate of Compliance.