Learn More
MSC GF Gage Threaded Ring Gage, 3/8-24 Thread, UNF, Class 2A, No Go
Catalog No. 5000515029
Encompass
This threaded ring gage is designed for inspecting 3/8-24 UNF threads. It features a Class 2A classification and is specifically a No Go gage, used to ensure that the thread does not exceed the maximum material limit.
  • No Go functionality for precise thread inspection.
  • Designed for 3/8-24 thread size.
  • Compatible with UNF thread types.
  • Ensures Class 2A thread accuracy.
  • Single ring design.
Catalog No. 50-005-15029 Supplier MSC Supplier No. 55323117
This threaded ring gage is designed for inspecting 3/8-24 UNF threads. It features a Class 2A classification and is specifically a No Go gage, used to ensure that the thread does not exceed the maximum material limit.
  • No Go functionality for precise thread inspection.
  • Designed for 3/8-24 thread size.
  • Compatible with UNF thread types.
  • Ensures Class 2A thread accuracy.
  • Single ring design.