Learn More
MSC GF Gage Threaded Ring Gage, 3/8-32 UNEF, 3A, Go/No Go
Catalog No. 5000528820
Encompass
This threaded ring gage is used for accurately measuring threads. It features a double-ring design and is specifically designed for 3/8-32 UNEF threads with a Class 3A classification, including both Go and No Go gauges.
  • Features Go and No Go measurement capabilities.
  • Designed for 3/8-32 thread size.
  • Supports UNEF thread type.
  • Meets Class 3A classification.
  • Features a double ring design.
  • Manufactured in the United States.
Catalog No. 50-005-28820 Supplier MSC Supplier No. 55326615
This threaded ring gage is used for accurately measuring threads. It features a double-ring design and is specifically designed for 3/8-32 UNEF threads with a Class 3A classification, including both Go and No Go gauges.
  • Features Go and No Go measurement capabilities.
  • Designed for 3/8-32 thread size.
  • Supports UNEF thread type.
  • Meets Class 3A classification.
  • Features a double ring design.
  • Manufactured in the United States.