Learn More
MSC GF Gage Threaded Ring Gage, #4-40, UNC, Class 2A, Go
Catalog No. 5000530184
Encompass
This threaded ring gage is designed for precise measurement of screw threads. It is specifically made for #4-40 UNC threads with a Class 2A classification and functions as a 'Go' gage, ensuring that external threads meet minimum material limits. This single ring type gage is constructed from tool steel, providing durability and accuracy for quality control applications.
  • Functions as a go gage
  • Designed for #4-40 thread size
  • Features UNC thread type
  • Includes 2A classification
  • Single ring design
  • Made from tool steel
Catalog No. 50-005-30184 Supplier MSC Supplier No. 55320410
This threaded ring gage is designed for precise measurement of screw threads. It is specifically made for #4-40 UNC threads with a Class 2A classification and functions as a 'Go' gage, ensuring that external threads meet minimum material limits. This single ring type gage is constructed from tool steel, providing durability and accuracy for quality control applications.
  • Functions as a go gage
  • Designed for #4-40 thread size
  • Features UNC thread type
  • Includes 2A classification
  • Single ring design
  • Made from tool steel