Learn More
MSC GF Gage Threaded Ring Gage, #4-48 Thread, Class 2A, No Go
Catalog No. 5000515028
Encompass
This GF Gage threaded ring gage is designed for precise measurement of external threads. It features a #4-48 thread size with a Class 2A tolerance, specifically configured as a 'No Go' gage to verify the maximum material limit of the thread.
  • Ensures accurate inspection of external threads.
  • Verifies maximum material limit of #4-48 Class 2A threads.
  • Provides reliable quality control for manufacturing processes.
  • Helps prevent assembly issues by confirming thread specifications.
Catalog No. 50-005-15028 Supplier MSC Supplier No. 55322754
This GF Gage threaded ring gage is designed for precise measurement of external threads. It features a #4-48 thread size with a Class 2A tolerance, specifically configured as a 'No Go' gage to verify the maximum material limit of the thread.
  • Ensures accurate inspection of external threads.
  • Verifies maximum material limit of #4-48 Class 2A threads.
  • Provides reliable quality control for manufacturing processes.
  • Helps prevent assembly issues by confirming thread specifications.