Learn More
MSC GF Gage Threaded Ring Gage, #5-40 Thread, Class 2A, No Go
Catalog No. 5000536864
Encompass
This threaded ring gage is designed for inspecting internal threads with a #5-40 thread size and 2A classification. It functions as a 'No Go' gage, ensuring the maximum material limit of the thread is not exceeded. Its single-ring design provides straightforward application for precise thread verification.
  • Designed for #5-40 thread inspection
  • Features a 2A classification
  • Functions as a No Go gage for maximum material limit verification
  • Single ring type for ease of use
  • Manufactured in the United States
Catalog No. 50-005-36864 Supplier MSC Supplier No. 55322770
This threaded ring gage is designed for inspecting internal threads with a #5-40 thread size and 2A classification. It functions as a 'No Go' gage, ensuring the maximum material limit of the thread is not exceeded. Its single-ring design provides straightforward application for precise thread verification.
  • Designed for #5-40 thread inspection
  • Features a 2A classification
  • Functions as a No Go gage for maximum material limit verification
  • Single ring type for ease of use
  • Manufactured in the United States