Learn More
MSC GF Gage Threaded Ring Gage, 5/8-24, UNEF, Class 2A, No Go
Catalog No. 5000531614
Encompass
This threaded ring gage is designed for precise measurement of 5/8-24 UNEF threads with a Class 2A classification. It functions as a No Go gage to ensure dimensional accuracy.
  • Functions as a no go threaded ring gage
  • Designed for 5/8-24 UNEF threads
  • Features a 2A thread classification
  • Single ring design
Catalog No. 50-005-31614 Supplier MSC Supplier No. 55323372
This threaded ring gage is designed for precise measurement of 5/8-24 UNEF threads with a Class 2A classification. It functions as a No Go gage to ensure dimensional accuracy.
  • Functions as a no go threaded ring gage
  • Designed for 5/8-24 UNEF threads
  • Features a 2A thread classification
  • Single ring design