Learn More
MSC GF Gage Threaded Ring Gage, 7/16-20 Thread, UNF, Class 2A, No Go
Catalog No. 5000527416
Encompass
This threaded ring gage is designed for precise measurement applications, featuring a 7/16-20 UNF thread with a Class 2A classification. It functions as a 'No Go' gage, ensuring components meet specific tolerance requirements for external threads. This single ring gage provides accuracy and reliability for quality control.
  • Functions as a 'No Go' gage
  • Features a 7/16-20 thread size
  • UNF thread type
  • Class 2A classification
  • Single ring design
  • Manufactured in the United States
Catalog No. 50-005-27416 Supplier MSC Supplier No. 55323174
This threaded ring gage is designed for precise measurement applications, featuring a 7/16-20 UNF thread with a Class 2A classification. It functions as a 'No Go' gage, ensuring components meet specific tolerance requirements for external threads. This single ring gage provides accuracy and reliability for quality control.
  • Functions as a 'No Go' gage
  • Features a 7/16-20 thread size
  • UNF thread type
  • Class 2A classification
  • Single ring design
  • Manufactured in the United States