Learn More
MSC GF Gage Threaded Ring Gage, 7/8-14 Thread, UNF, Class 3A, Go
Catalog No. 5000527455
Encompass
This threaded ring gage is designed for precise measurement of 7/8-14 UNF 3A 'Go' threads. It ensures compliance with specified thread tolerances, making it suitable for quality control and inspection applications.
  • Designed for 7/8-14 thread size
  • Supports UNF thread type
  • Features 3A classification for accuracy
  • "Go" designation for checking maximum material limits
  • Single ring configuration
Catalog No. 50-005-27455 Supplier MSC Supplier No. 55322317
This threaded ring gage is designed for precise measurement of 7/8-14 UNF 3A 'Go' threads. It ensures compliance with specified thread tolerances, making it suitable for quality control and inspection applications.
  • Designed for 7/8-14 thread size
  • Supports UNF thread type
  • Features 3A classification for accuracy
  • "Go" designation for checking maximum material limits
  • Single ring configuration