Learn More
MSC GF Gage Threaded Ring Gage, 7/8-20 Thread, UNEF, Class 2A, Go
Catalog No. 5000528215
Encompass
This threaded ring gage is designed for inspecting 7/8-20 UNEF threads with a Class 2A classification. It serves as a "Go" gage to ensure the manufactured thread is not too large and will accept the mating part.
  • Go gage for thread inspection
  • 7/8-20 thread size
  • UNEF thread type
  • Class 2A classification
  • Single ring design
  • Manufactured in the United States
Catalog No. 50-005-28215 Supplier MSC Supplier No. 55321178
This threaded ring gage is designed for inspecting 7/8-20 UNEF threads with a Class 2A classification. It serves as a "Go" gage to ensure the manufactured thread is not too large and will accept the mating part.
  • Go gage for thread inspection
  • 7/8-20 thread size
  • UNEF thread type
  • Class 2A classification
  • Single ring design
  • Manufactured in the United States