Learn More
MSC GF Gage Threaded Ring Gage, 7/8-20 Thread, UNEF, Class 3A, Go & No Go
Catalog No. 5000514477
Encompass
This threaded ring gage is designed for precise measurement and verification of external threads. It features a dual Go & No Go design to ensure threaded components meet specified maximum and minimum material limits, ensuring proper fit and functionality.
  • Checks both maximum and minimum material limits.
  • Designed for 7/8-20 UNEF threads with Class 3A classification.
  • Double ring configuration for comprehensive inspection.
  • Manufactured in the United States.
Catalog No. 50-005-14477 Supplier MSC Supplier No. 55326979
This threaded ring gage is designed for precise measurement and verification of external threads. It features a dual Go & No Go design to ensure threaded components meet specified maximum and minimum material limits, ensuring proper fit and functionality.
  • Checks both maximum and minimum material limits.
  • Designed for 7/8-20 UNEF threads with Class 3A classification.
  • Double ring configuration for comprehensive inspection.
  • Manufactured in the United States.