Learn More
MSC GF Gage Threaded Ring Gage, M12 x 1.75, 6G, Go/No Go
Catalog No. 5000527459
Encompass
This threaded ring gage is designed for inspecting M12 x 1.75 metric threads. It includes both Go and No Go features to ensure precise thread measurement and conformity. Supplied as a 2-piece set, this gage is suitable for quality control and manufacturing applications.
  • Go and No Go design for comprehensive thread inspection.
  • M12 x 1.75 thread size.
  • Class 6G classification.
  • Supplied as a 2 piece set.
  • Includes a certificate of conformance.
  • Manufactured in the United States.
Catalog No. 50-005-27459 Supplier MSC Supplier No. 55335095
This threaded ring gage is designed for inspecting M12 x 1.75 metric threads. It includes both Go and No Go features to ensure precise thread measurement and conformity. Supplied as a 2-piece set, this gage is suitable for quality control and manufacturing applications.
  • Go and No Go design for comprehensive thread inspection.
  • M12 x 1.75 thread size.
  • Class 6G classification.
  • Supplied as a 2 piece set.
  • Includes a certificate of conformance.
  • Manufactured in the United States.