Learn More
MSC GF Gage Threaded Ring Gage, M14 x 1.50 Thread, Metric, Class 6G, Go & No Go
Catalog No. 5000532041
Encompass
This Threaded Ring Gage is designed for precise measurement of M14 x 1.50 Metric threads. It features both "Go" and "No Go" gaging capabilities, ensuring accurate verification of thread integrity according to Class 6G specifications.
  • Designed for M14 x 1.50 Metric threads.
  • Includes both "Go" and "No Go" gaging for comprehensive thread verification.
  • Class 6G classification ensures adherence to specific quality standards.
  • Double ring design.
  • Manufactured in the United States.
Catalog No. 50-005-32041 Supplier MSC Supplier No. 55335152
This Threaded Ring Gage is designed for precise measurement of M14 x 1.50 Metric threads. It features both "Go" and "No Go" gaging capabilities, ensuring accurate verification of thread integrity according to Class 6G specifications.
  • Designed for M14 x 1.50 Metric threads.
  • Includes both "Go" and "No Go" gaging for comprehensive thread verification.
  • Class 6G classification ensures adherence to specific quality standards.
  • Double ring design.
  • Manufactured in the United States.