Learn More
MSC GF Gage Threaded Ring Gage, M18 x 1.50 Thread, Metric, Class 6G, Go & No Go
Catalog No. 5000527648
Encompass
This high-precision metric Go & No Go gauge is designed for accurate thread verification. Featuring a Class 6G classification, this double-ring set ensures precise measurement of M18x1.5 threads.
  • Ensures accurate thread measurement with Go & No Go functionality.
  • Metric M18x1.5 thread size for specific applications.
  • Class 6G classification guarantees high precision.
  • Double ring design for comprehensive inspection.
  • Manufactured in the United States.
Catalog No. 50-005-27648 Supplier MSC Supplier No. 55335210
This high-precision metric Go & No Go gauge is designed for accurate thread verification. Featuring a Class 6G classification, this double-ring set ensures precise measurement of M18x1.5 threads.
  • Ensures accurate thread measurement with Go & No Go functionality.
  • Metric M18x1.5 thread size for specific applications.
  • Class 6G classification guarantees high precision.
  • Double ring design for comprehensive inspection.
  • Manufactured in the United States.