Learn More
MSC GF Gage Threaded Ring Gage, M6 x 1.00 Metric, Class 6G, No Go
Catalog No. 5000530189
Encompass
This threaded ring gage is designed for checking M6 x 1.00 metric threads. It is a 'No Go' gage, ensuring the part does not exceed the maximum material limit. It features a Class 6G classification for precise measurement.
  • No Go functionality for quality control
  • M6 x 1.00 metric thread size
  • Single ring design
  • Class 6G classification for accuracy
  • Made in the United States
Catalog No. 50-005-30189 Supplier MSC Supplier No. 55334650
This threaded ring gage is designed for checking M6 x 1.00 metric threads. It is a 'No Go' gage, ensuring the part does not exceed the maximum material limit. It features a Class 6G classification for precise measurement.
  • No Go functionality for quality control
  • M6 x 1.00 metric thread size
  • Single ring design
  • Class 6G classification for accuracy
  • Made in the United States