Learn More
MSC GF Gage Threaded Ring Gage, M8 x 1.25 Metric, Class 6G, Go
Catalog No. 5000528795
Encompass
This threaded ring gage is designed for accurately checking external thread dimensions. It features an M8 x 1.25 metric thread and is classified as a Class 6G 'Go' gage, ensuring precise and reliable measurement for quality control applications.
  • Accurately checks external thread dimensions
  • M8 x 1.25 thread size
  • Metric thread type
  • Class 6G classification
  • "Go" type gage for acceptance testing
  • Single ring design
  • Manufactured in the United States
Catalog No. 50-005-28795 Supplier MSC Supplier No. 55334312
This threaded ring gage is designed for accurately checking external thread dimensions. It features an M8 x 1.25 metric thread and is classified as a Class 6G 'Go' gage, ensuring precise and reliable measurement for quality control applications.
  • Accurately checks external thread dimensions
  • M8 x 1.25 thread size
  • Metric thread type
  • Class 6G classification
  • "Go" type gage for acceptance testing
  • Single ring design
  • Manufactured in the United States