Learn More
MSC GF Gage Threaded Ring Gage, No Go, 7/8-20, UNEF, Class 2A
Catalog No. 5000527413
Encompass
This threaded ring gage is designed for precise inspection of external threads, specifically to ensure that the threads are not oversized. It is a 'No Go' gage, used to confirm that the thread's maximum material limit is not exceeded, helping maintain manufacturing quality and part interchangeability.
  • No go functionality for checking maximum thread limits
  • Designed for 7/8-20 thread size
  • Features UNEF thread type
  • Class 2A classification for specific thread fit
  • Single ring design
  • Manufactured in the United States
Catalog No. 50-005-27413 Supplier MSC Supplier No. 55323497
This threaded ring gage is designed for precise inspection of external threads, specifically to ensure that the threads are not oversized. It is a 'No Go' gage, used to confirm that the thread's maximum material limit is not exceeded, helping maintain manufacturing quality and part interchangeability.
  • No go functionality for checking maximum thread limits
  • Designed for 7/8-20 thread size
  • Features UNEF thread type
  • Class 2A classification for specific thread fit
  • Single ring design
  • Manufactured in the United States