Learn More
MSC GF Gage Truncated Taperlock Gage, 1-1/4-12 UNF, Go & No Go, Class 3A
Catalog No. 5000528794
Encompass
This precision thread gage is a Truncated Taperlock Gage designed for 1-1/4-12 UNF threads. It features both 'Go' and 'No Go' ends to ensure accurate thread inspection, built to Class 3A tolerance standards.
  • Designed for 1-1/4-12 UNF thread inspection
  • Includes both Go and No Go ends for comprehensive thread assessment
  • Manufactured to Class 3A tolerance standards
  • Equipped with a 5 in handle for ease of use
Catalog No. 50-005-28794 Supplier MSC Supplier No. 55331771
This precision thread gage is a Truncated Taperlock Gage designed for 1-1/4-12 UNF threads. It features both 'Go' and 'No Go' ends to ensure accurate thread inspection, built to Class 3A tolerance standards.
  • Designed for 1-1/4-12 UNF thread inspection
  • Includes both Go and No Go ends for comprehensive thread assessment
  • Manufactured to Class 3A tolerance standards
  • Equipped with a 5 in handle for ease of use