Learn More
MSC GF Gage Truncated Taperlock Gage, 1-8 UNC, Class 3A, No Go
Catalog No. 5000531242
Encompass
This GF Gage Truncated Taperlock Gage is designed for precision thread measurement. It features a 1-8 UNC thread size and is specified for Class 3A applications as a No Go gage.
  • Designed for precision thread measurement
  • Features 1-8 UNC thread size
  • Suitable for Class 3A applications
  • No Go configuration for inspection
Catalog No. 50-005-31242 Supplier MSC Supplier No. 55331631
This GF Gage Truncated Taperlock Gage is designed for precision thread measurement. It features a 1-8 UNC thread size and is specified for Class 3A applications as a No Go gage.
  • Designed for precision thread measurement
  • Features 1-8 UNC thread size
  • Suitable for Class 3A applications
  • No Go configuration for inspection