Learn More
MSC GF Gage Truncated Taperlock Gage, #10-32 UNF, No Go, Class 2A, Steel
Catalog No. 5000531100
Encompass
The GF Gage truncated taperlock gage is a no go thread plug gage designed for precisely checking #10-32 UNF threads with a 2A classification. Constructed from durable steel, it ensures accurate measurement in various applications.
  • Truncated taperlock thread gage type
  • Designed for #10-32 thread size
  • No Go functionality for thread inspection
  • Features a 2A thread classification
  • Suitable for UNF thread types
  • Made from high-quality steel for durability
Catalog No. 50-005-31100 Supplier MSC Supplier No. 55329874
The GF Gage truncated taperlock gage is a no go thread plug gage designed for precisely checking #10-32 UNF threads with a 2A classification. Constructed from durable steel, it ensures accurate measurement in various applications.
  • Truncated taperlock thread gage type
  • Designed for #10-32 thread size
  • No Go functionality for thread inspection
  • Features a 2A thread classification
  • Suitable for UNF thread types
  • Made from high-quality steel for durability