Learn More
MSC GF Gage Truncated Taperlock Gage, 3/4-16, Go & No Go, 3A
Catalog No. 5000535849
Encompass
This truncated taperlock thread gage is designed for precise measurement and inspection of internal threads. It features both 'Go' and 'No Go' sides, ensuring accuracy for 3/4-16 size threads with a Class 3A fit. Constructed from durable steel, it is suitable for various industrial applications requiring thread verification.
  • Truncated taperlock thread gage type
  • Designed for 3/4-16 thread size
  • Includes both go and no go sides for accurate measurement
  • Features a class 3A thread classification
  • Constructed from steel for durability
Catalog No. 50-005-35849 Supplier MSC Supplier No. 55333850
This truncated taperlock thread gage is designed for precise measurement and inspection of internal threads. It features both 'Go' and 'No Go' sides, ensuring accuracy for 3/4-16 size threads with a Class 3A fit. Constructed from durable steel, it is suitable for various industrial applications requiring thread verification.
  • Truncated taperlock thread gage type
  • Designed for 3/4-16 thread size
  • Includes both go and no go sides for accurate measurement
  • Features a class 3A thread classification
  • Constructed from steel for durability