Learn More
MSC GF Gage Truncated Taperlock Gage, M6 x 1.00, Go, Class 6G, 1 in Handle
Catalog No. 5000537447
Encompass
This truncated taperlock gage is designed for M6 x 1.00 threads. It is a "Go" gage, featuring a Class 6G classification and a 1-inch handle for ease of use.
  • Truncated taperlock thread gage type.
  • M6 x 1.00 thread size.
  • Go condition for thread verification.
  • Class 6G classification.
  • Includes a 1-inch handle.
  • Country of origin: United States.
Catalog No. 50-005-37447 Supplier MSC Supplier No. 55335376
This truncated taperlock gage is designed for M6 x 1.00 threads. It is a "Go" gage, featuring a Class 6G classification and a 1-inch handle for ease of use.
  • Truncated taperlock thread gage type.
  • M6 x 1.00 thread size.
  • Go condition for thread verification.
  • Class 6G classification.
  • Includes a 1-inch handle.
  • Country of origin: United States.