Learn More
MSC GF Gage Truncated Taperlock Thread Gage, 1-1/8-12, Go & No Go, Class 2A
Catalog No. 5000527720
Encompass
GF Gage Truncated Taperlock Gages are designed for inspecting internal threads. This specific gage features a 1-1/8-12 thread size and is classified as 2A, providing both Go and No Go measurements for comprehensive thread verification.
  • Truncated Taperlock thread gage type
  • 1-1/8-12 thread size
  • 4 in handle size
  • Includes both Go and No Go measurements
  • Class 2A classification
  • Constructed from steel material
Catalog No. 50-005-27720 Supplier MSC Supplier No. 55332894
GF Gage Truncated Taperlock Gages are designed for inspecting internal threads. This specific gage features a 1-1/8-12 thread size and is classified as 2A, providing both Go and No Go measurements for comprehensive thread verification.
  • Truncated Taperlock thread gage type
  • 1-1/8-12 thread size
  • 4 in handle size
  • Includes both Go and No Go measurements
  • Class 2A classification
  • Constructed from steel material