Learn More
MSC GF Gage Truncated Taperlock Thread Gage, 1/4-32 UNEF, Class 2A, Steel
Catalog No. 5000514741
Encompass
This Truncated Taperlock Thread Gage is designed for precise measurement of 1/4-32 UNEF threads. It features both "Go" and "No Go" functions to ensure accurate thread inspection and is classified as 2A. Made from durable steel, this gage includes a 1-inch handle for ease of use.
  • Truncated taperlock thread gage type
  • Thread size of 1/4-32
  • Features both Go and No Go functions
  • Class 2A for precise measurement
  • Constructed from durable steel
  • Includes a 1-inch handle
Catalog No. 50-005-14741 Supplier MSC Supplier No. 55332290
This Truncated Taperlock Thread Gage is designed for precise measurement of 1/4-32 UNEF threads. It features both "Go" and "No Go" functions to ensure accurate thread inspection and is classified as 2A. Made from durable steel, this gage includes a 1-inch handle for ease of use.
  • Truncated taperlock thread gage type
  • Thread size of 1/4-32
  • Features both Go and No Go functions
  • Class 2A for precise measurement
  • Constructed from durable steel
  • Includes a 1-inch handle