Learn More
MSC GF Gage Truncated Taperlock Thread Gage, #4-40 UNC, Go/No Go, Class 2A
Catalog No. 5000511505
Encompass
This Truncated Taperlock Thread Gage Set includes both Go and No Go gages for #4-40 UNC threads, featuring a 2A classification. Constructed from steel, it is designed for precise thread measurement and comes equipped with a handle for ease of use.
  • Truncated Taperlock thread gage type
  • For #4-40 thread size
  • Includes both Go and No Go gages
  • 2A classification
  • Made of steel
  • Includes a handle
Catalog No. 50-005-11505 Supplier MSC Supplier No. 55332019
This Truncated Taperlock Thread Gage Set includes both Go and No Go gages for #4-40 UNC threads, featuring a 2A classification. Constructed from steel, it is designed for precise thread measurement and comes equipped with a handle for ease of use.
  • Truncated Taperlock thread gage type
  • For #4-40 thread size
  • Includes both Go and No Go gages
  • 2A classification
  • Made of steel
  • Includes a handle