Learn More
MSC GF Gage Truncated Taperlock Thread Gage, #5-44, No Go, Class 3A
Catalog No. 5000529000
Encompass
This Truncated Taperlock Thread Gage is designed for inspecting #5-44 threads, specifically functioning as a "No Go" gage with a Class 3A classification. It is constructed from steel, ensuring durability and precision.
  • Truncated Taperlock thread gage type
  • #5-44 thread size
  • No Go functionality
  • 3A classification
  • Steel construction
  • Manufactured in the United States
Catalog No. 50-005-29000 Supplier MSC Supplier No. 55330914
This Truncated Taperlock Thread Gage is designed for inspecting #5-44 threads, specifically functioning as a "No Go" gage with a Class 3A classification. It is constructed from steel, ensuring durability and precision.
  • Truncated Taperlock thread gage type
  • #5-44 thread size
  • No Go functionality
  • 3A classification
  • Steel construction
  • Manufactured in the United States