Learn More
MSC GF Gage Truncated Taperlock Thread Gage, 5/8-24 Thread Size, Go, Class 2A
Catalog No. 5000535843
Encompass
This Truncated Taperlock Thread Gage provides precise measurement for industrial applications. It features a 5/8-24 thread size and a 'Go' classification of 2A, ensuring accurate thread inspection. Constructed from durable steel, this gage is built for longevity and reliable performance.
  • Truncated Taperlock Thread Gage type
  • 5/8-24 Thread size
  • 3 inch handle for comfortable grip
  • Class 2A classification for accuracy
  • 'Go' designation for quick pass/fail inspection
  • Durable steel construction
Catalog No. 50-005-35843 Supplier MSC Supplier No. 55328017
This Truncated Taperlock Thread Gage provides precise measurement for industrial applications. It features a 5/8-24 thread size and a 'Go' classification of 2A, ensuring accurate thread inspection. Constructed from durable steel, this gage is built for longevity and reliable performance.
  • Truncated Taperlock Thread Gage type
  • 5/8-24 Thread size
  • 3 inch handle for comfortable grip
  • Class 2A classification for accuracy
  • 'Go' designation for quick pass/fail inspection
  • Durable steel construction