Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graham-Field™ Grafco™ Disposable Penlites
SDP

Catalog No. 19027480
Change view
Click to view available options
Product Type:
Disposable Diagnostic Penlight
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-027480 Disposable Diagnostic Penlight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-027480 Supplier Graham-Field™ Supplier No. 4062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compact, lightweight and ready to use

Deliver high-intensity bright light without the inconvenience of changing batteries or bulbs.

  • Ideal for the physician, nurse, technician or paramedic
  • Bright-white, low-drain LED
  • Latex-free
  • Shelf life of one year

Specifications

Light Source Low-drain LED
Color Bright White
For Use With (Application) Compact, lightweight and ready to use, deliver high-intensity bright light without the inconvenience of changing batteries or bulbs, for the physician, nurse, technician or paramedic
Product Type Disposable Diagnostic Penlight
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.