Learn More
Cole-Parmer GF Signet 3-2725-60 ORP Electrode, Flat Surface, 3/4" NPT Threads
Catalog No. NC1589853 Shop All Cole-Parmer Products
Encompass
Electrodes use a unique foul-proof DryLoc connector with gold plated contacts for extended lifetime. Choice of flat pH, bulb pH, and flat ORP sensing elements provides for a broad range of uses. HF resistant versions are for use in applications where traces of hydrofluoric acid will attack standard glass. LC versions are designed for use in low conductivity fluids and ultrapure water applications.
Catalog No. NC1589853 Supplier Cole-Parmer Supplier No. 5656116
Electrodes use a unique foul-proof DryLoc connector with gold plated contacts for extended lifetime. Choice of flat pH, bulb pH, and flat ORP sensing elements provides for a broad range of uses. HF resistant versions are for use in applications where traces of hydrofluoric acid will attack standard glass. LC versions are designed for use in low conductivity fluids and ultrapure water applications.