Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAA Instrument Calibration Standard 1 (CLP), Ricca Chemical
SDP

Catalog No. RGINSCL150 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RGINSCL1-50 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RGINSCL1-50 Supplier Ricca Chemical Company Supplier No. RGINSCL150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material GFAA Instrument Calibration Standard
Packaging Natural Poly Bottle
CAS 7664-39-3, 7732-18-5, 1309-64-4, 7782-49-2, 7439-92-1
CAS Min % 91.0, 7.45, 0.01, 0.01, 0.01, 0.0043
CAS Max % 94.0, 7.76, 0.01, 0.01, 0.01, 0.0045
Odor Odorless
Color Colorless
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 50 mL
Grade CLP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.