Learn More
Diagnostic Biosystems GFAP Glial Fibrillary Acidic Protein, GFAP
Catalog No. 502407395
Encompass_Preferred
This antibody reacts with human GFAP. In the central nervous system, this antibody stains astrocytes and some groups of ependymal cells. However, in the peripheral nervous system, it stains Schwann cells, satellite cells and enteric glial cells.
Catalog No. 50-240-7395 Supplier Diagnostic Biosystems Supplier No. PDR028
This antibody reacts with human GFAP. In the central nervous system, this antibody stains astrocytes and some groups of ependymal cells. However, in the peripheral nervous system, it stains Schwann cells, satellite cells and enteric glial cells.