Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2670MSM1XXP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM507. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Brain or Astrocytoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: GFAP isolated from pig spinal cord. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~50kDa.Chromosome: 17q21.31. Gene Symbol: GFAP. Gene ID: 2670. Ensemble: ENSG00000131095. Uniprot: NP_002046.
Catalog No. 50-188-4730