Learn More
Aviva Systems Biology Corp Gfap Antibody - N-terminal region (ARP54621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gfap. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI836096ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-947