Learn More
Biomatik Corporation GFAP Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50131
Encompass_Preferred
GFAP Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Glial Fibrillary Acidic Protein. The GFAP Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 292~432aa. Applications: ELISA, WB, IHC, IF. Target Synonyms: Glial Fibrillary Acidic Protein. Accession Number: P14136. Gene ID: 2670. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7383