Learn More
Diagnostic Biosystems GFAP CONC. POLYCLONAL AB 1ML Glial Fibrillary Acidic Protein, GFAP
Supplier:  Diagnostic Biosystems RP014
Encompass_Preferred
This antibody reacts with human GFAP. In the central nervous system, this antibody stains astrocytes and some groups of ependymal cells. However, in the peripheral nervous system, it stains Schwann cells, satellite cells and enteric glial cells.
Catalog No. NC1816723