Learn More
Cayman Chemical GFAP MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502941077
Encompass_Preferred
Immunogen: Full length recombinant human GFAP protein • Host: Mouse • Species Reactivity: (+) Human, mouse • Applications: ELISA, IHC, and WB • MW: ~50 kDa
Catalog No. 50-294-1077 Supplier Cayman Chemical Supplier No. 28847100 UG
Immunogen: Full length recombinant human GFAP protein • Host: Mouse • Species Reactivity: (+) Human, mouse • Applications: ELISA, IHC, and WB • MW: ~50 kDa