Learn More
Medchemexpress LLC GFER Human N-His 10ug
SDP
Catalog No. 5000197040
Encompass_Preferred
Recombinant human GFER protein with an N-terminal His tag, expressed in E. coli and supplied lyophilized for research use. The protein is 125 amino acids (~15 kDa), with >95% purity by reducing SDS-PAGE, and is formulated in 50 mM Glycine-HCl, 150 mM NaCl (pH 2.5). It is suitable for studies of liver regeneration and mitochondrial disulfide relay; reconstitute to ≥100 μg/mL and aliquot to avoid freeze-thaw degradation.
Catalog No. 50-001-97040 Supplier Medchemexpress LLC Supplier No. HYP70953A10UG
Recombinant human GFER protein with an N-terminal His tag, expressed in E. coli and supplied lyophilized for research use. The protein is 125 amino acids (~15 kDa), with >95% purity by reducing SDS-PAGE, and is formulated in 50 mM Glycine-HCl, 150 mM NaCl (pH 2.5). It is suitable for studies of liver regeneration and mitochondrial disulfide relay; reconstitute to ≥100 μg/mL and aliquot to avoid freeze-thaw degradation.