Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GFER Human N-His 50ug
SDP

Catalog No. 5000197101
Encompass_Preferred

GFER Protein, Human (N-His) is a recombinant human growth factor supplied lyophilized with an N-terminal 6xHis tag and expressed in E. coli. The preparation is intended for biochemical and cell-based research related to protein folding and liver regeneration, provided at high purity for assay development and interaction studies.

  • Expressed in E. coli with an N-terminal His tag for easy purification.
  • Lyophilized formulation for long-term storage and convenient transport.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximately 15 kDa molecular weight suitable for biochemical assays.
  • Recommended storage at -20°C or -80°C with aliquots to preserve activity.
  • Includes sequence information to support assay design and antibody selection.

Catalog No. 50-001-97101 Supplier Medchemexpress LLC Supplier No. HYP70953A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GFER Protein, Human (N-His) is a recombinant human growth factor supplied lyophilized with an N-terminal 6xHis tag and expressed in E. coli. The preparation is intended for biochemical and cell-based research related to protein folding and liver regeneration, provided at high purity for assay development and interaction studies.

  • Expressed in E. coli with an N-terminal His tag for easy purification.
  • Lyophilized formulation for long-term storage and convenient transport.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximately 15 kDa molecular weight suitable for biochemical assays.
  • Recommended storage at -20°C or -80°C with aliquots to preserve activity.
  • Includes sequence information to support assay design and antibody selection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.