Learn More
Aviva Systems Biology Corp GFI1B Antibody - N-terminal region (ARP30093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFI1B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GFI1B. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54122