Learn More
MSC GF Gage Threaded Ring Gage, M16 x 2.00, Metric, 6G, Go, 1 ea.
Catalog No. 5000514744
Encompass
This metric threaded ring gage is designed for precise inspection and verification of external threads. It features a thread size of M16 x 2.00 and is manufactured to a 6G classification, specifically for Go applications.
  • Metric thread size of M16 x 2.00
  • Class 6G classification
  • Single Go ring gage design
  • Suitable for professional measurement and inspection tasks
Catalog No. 50-005-14744 Supplier MSC Supplier No. 55334452
This metric threaded ring gage is designed for precise inspection and verification of external threads. It features a thread size of M16 x 2.00 and is manufactured to a 6G classification, specifically for Go applications.
  • Metric thread size of M16 x 2.00
  • Class 6G classification
  • Single Go ring gage design
  • Suitable for professional measurement and inspection tasks