Learn More
MSC GF Gage Threaded Ring Gage, M18 x 1.5, 6G, No Go, 1 ea
Catalog No. 5000514510
Encompass
This metric thread ring gage is designed for inspecting the external thread profile of parts. It is a single, No Go gage used to ensure that manufactured threads do not exceed specified tolerances.
  • Metric thread type
  • M18 x 1.50 thread size
  • Class 6G classification
  • No Go configuration
Catalog No. 50-005-14510 Supplier MSC Supplier No. 55334858
This metric thread ring gage is designed for inspecting the external thread profile of parts. It is a single, No Go gage used to ensure that manufactured threads do not exceed specified tolerances.
  • Metric thread type
  • M18 x 1.50 thread size
  • Class 6G classification
  • No Go configuration