Learn More
MSC Gf Gage Pipe Thread Plug Gage, Tapered, 1-1/2-11-1/2, Class L-1, Single End, 1 ea
Catalog No. 5000511553
Encompass
This tapered pipe thread plug gage is designed for precision inspection of NPT-L1 threads. It features a single end configuration for reliable go/no-go verification in metalworking and manufacturing applications.
  • Thread size of 1-1/2-11-1/2
  • Class L-1 tolerance
  • Tapered thread geometry
  • Single end design
  • Handle size 5
Catalog No. 50-005-11553 Supplier MSC Supplier No. 55336515
This tapered pipe thread plug gage is designed for precision inspection of NPT-L1 threads. It features a single end configuration for reliable go/no-go verification in metalworking and manufacturing applications.
  • Thread size of 1-1/2-11-1/2
  • Class L-1 tolerance
  • Tapered thread geometry
  • Single end design
  • Handle size 5