Learn More
MSC GF Gage Threaded Pipe Ring, NPTF, 3/4-14, 6 Step, 1 ea.
Catalog No. 5000514556
Encompass
This threaded pipe ring is a specialized gauging tool designed for precision measurement applications. Engineered with a 6 step classification, this instrument is suitable for evaluating NPTF threads with a size of 3/4-14.
  • Class 6 step design for accurate thread assessment
  • Compatible with 3/4-14 thread sizes
  • NPTF thread type
Catalog No. 50-005-14556 Supplier MSC Supplier No. 55337737
This threaded pipe ring is a specialized gauging tool designed for precision measurement applications. Engineered with a 6 step classification, this instrument is suitable for evaluating NPTF threads with a size of 3/4-14.
  • Class 6 step design for accurate thread assessment
  • Compatible with 3/4-14 thread sizes
  • NPTF thread type