Learn More
MSC GF Gage Pipe Thread Plug Gage, 1-11-1/2 NPTF-L1, Tapered, Single End, 1 ea
Catalog No. 5000547936
Encompass
This single end tapered pipe thread plug gage is designed for precise inspection of NPTF-L1 threads. It is engineered to meet rigorous standards for checking thread accuracy and consistency in industrial applications.
  • Tapered design for accurate pipe thread inspection
  • Size specification of 1-11-1/2
  • Single end configuration
  • Handle size 5
  • Manufactured in the United States
Catalog No. 50-005-47936 Supplier MSC Supplier No. 55337893
This single end tapered pipe thread plug gage is designed for precise inspection of NPTF-L1 threads. It is engineered to meet rigorous standards for checking thread accuracy and consistency in industrial applications.
  • Tapered design for accurate pipe thread inspection
  • Size specification of 1-11-1/2
  • Single end configuration
  • Handle size 5
  • Manufactured in the United States