Learn More
MSC GF Gage Truncated Taperlock Thread Gage, #8-32 UNC, 2A, Steel, 1 ea
Catalog No. 5000547968
Encompass
This truncated taperlock thread gage is designed for precise inspection of #8-32 UNC internal threads. The steel construction ensures durability and dimensional stability during repetitive use. This set includes both Go and No Go members to verify that threaded parts meet the required 2A classification standards.
  • Features a #8-32 UNC thread size
  • Designed with 2A classification
  • Constructed from durable steel
  • Includes handle for ease of operation
  • Provides both Go and No Go verification
Catalog No. 50-005-47968 Supplier MSC Supplier No. 55332134
This truncated taperlock thread gage is designed for precise inspection of #8-32 UNC internal threads. The steel construction ensures durability and dimensional stability during repetitive use. This set includes both Go and No Go members to verify that threaded parts meet the required 2A classification standards.
  • Features a #8-32 UNC thread size
  • Designed with 2A classification
  • Constructed from durable steel
  • Includes handle for ease of operation
  • Provides both Go and No Go verification