Learn More
MSC GF Gage Truncated Taperlock Gage, 1-1/8-7, Class 2A, 1 ea
Catalog No. 5000514806
Encompass
This truncated taperlock gage is designed for precise measurement of 1-1/8-7 thread specifications. It features a double-ended design, incorporating both Go and No Go members to ensure thread accuracy and quality control in industrial applications.
  • Class 2A tolerance
  • Double-ended configuration for Go and No Go testing
  • Includes a 4 in. handle for secure grip and ease of use
  • Truncated thread form
Catalog No. 50-005-14806 Supplier MSC Supplier No. 55332878
This truncated taperlock gage is designed for precise measurement of 1-1/8-7 thread specifications. It features a double-ended design, incorporating both Go and No Go members to ensure thread accuracy and quality control in industrial applications.
  • Class 2A tolerance
  • Double-ended configuration for Go and No Go testing
  • Includes a 4 in. handle for secure grip and ease of use
  • Truncated thread form