Learn More
MSC GF Gage Truncated Taperlock Thread Gage, 1-1/2-12, 3A, Steel, 1 ea
Catalog No. 5000511508
Encompass
This truncated taperlock thread gage is a high-precision tool designed for checking thread size and fit. It provides reliable measurements for 1-1/2-12 threads with a 3A classification, ensuring consistency in threaded components.
  • Constructed from durable steel for long-term wear resistance
  • Features a double-ended design for both go and no go verification
  • Designed with a size 5 handle for comfortable and secure operation
  • Engineered for accurate thread inspection in industrial applications
Catalog No. 50-005-11508 Supplier MSC Supplier No. 55334171
This truncated taperlock thread gage is a high-precision tool designed for checking thread size and fit. It provides reliable measurements for 1-1/2-12 threads with a 3A classification, ensuring consistency in threaded components.
  • Constructed from durable steel for long-term wear resistance
  • Features a double-ended design for both go and no go verification
  • Designed with a size 5 handle for comfortable and secure operation
  • Engineered for accurate thread inspection in industrial applications