Learn More
MSC GF Gage Truncated Taperlock Thread Plug Gage, 7/16-14, Class 3A, No Go, 1 ea
Catalog No. 5000512005
Encompass
This truncated Taperlock thread gage is designed for precision inspection of internal threads, specifically manufactured for the 7/16-14 thread size in class 3A. It serves as a No Go gauge to ensure components meet strict tolerance requirements in industrial metalworking applications.
  • Constructed from high-quality steel for durability and longevity
  • Features a No Go design for accurate thread verification
  • Utilizes a size 2 Taperlock handle configuration
  • Engineered for consistent and reliable thread inspection
Catalog No. 50-005-12005 Supplier MSC Supplier No. 55331276
This truncated Taperlock thread gage is designed for precision inspection of internal threads, specifically manufactured for the 7/16-14 thread size in class 3A. It serves as a No Go gauge to ensure components meet strict tolerance requirements in industrial metalworking applications.
  • Constructed from high-quality steel for durability and longevity
  • Features a No Go design for accurate thread verification
  • Utilizes a size 2 Taperlock handle configuration
  • Engineered for consistent and reliable thread inspection